Läs mer
L-Leucine-1-13C, TRC
Produktkod. 30478583
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478583 100 mg
30477302 1 g
30469075 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30478583 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL330625100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74292-94-7
Molekylformel C5 13C H13 N O2
Molekylvikt (g/mol) 132.17
Synonym L-[1-13C]Leucine, [1-13C]Leucine
IUPAC-namn (2S)-2-amino-4-methyl(113C)pentanoic acid
LEDER Cc(C)C[C@H](N)[13C](=O)O
Specifikationer
Kemiskt namn eller material L-Leucine-1-13C
InChI formel InChI=1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i6+1
Kvantitet 100 mg
Formel vikt 132.098