missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, MP Biomedicals

Catalog Number 11411901
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
61-90-5
C6H13NO2
131.18
MFCD00002617
ROHFNLRQFUQHCH-MLHKIVSYNA-N
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
6106
CHEBI:15603
(2S)-2-amino-4-methylpentanoic acid
CC(C)C[C@H](N)C(O)=O
This item is not returnable. View return policy
61-90-5
C6H13NO2
131.18
MFCD00002617
ROHFNLRQFUQHCH-MLHKIVSYNA-N
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
6106
CHEBI:15603
(2S)-2-amino-4-methylpentanoic acid
CC(C)C[C@H](N)C(O)=O
Formula Weight 131.2
Grade Cell Culture Reagent
Quantity 100 g
Physical Form Crystalline
Chemical Name or Material L-Leucine, Substantially free of isoleucine and methionine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'