Läs mer
L-Lysine Clonixinate, TRC
Produktkod. 17999444
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17999444 1 g
17958314 500 mg
17918014 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17999444 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4885301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55837-30-4
Molekylformel C13 H11 Cl N2 O2 . C6 H14 N2 O2
Molekylvikt (g/mol) 408.88
Synonym L-Lysine, 2-[(3-chloro-2-methylphenyl)amino]-3-pyridinecarboxylate (1:1), L-Lysine, mono[2-[(3-chloro-2-methylphenyl)amino]-3-pyridinecarboxylate] (9CI), 3-Pyridinecarboxylic acid, 2-[(3-chloro-2-methylphenyl)amino]-, compd. with L-lysine (1:1) (9CI), Clonix, Clonixin lysinate, Clonixin lysine salt, Deltar, Dolagial, Dorixina, L 104, L 104 (pharmaceutical), Lysine clonixinate
IUPAC-namn 2-(3-chloro-2-methylanilino)pyridine-3-carboxylic acid;(2S)-2,6-diaminohexanoic acid
LEDER Cc1c(Cl)cccc1Nc2ncccc2C(=O)O.NCCCC[C@H](N)C(=O)O
Specifikationer
Kemiskt namn eller material Lysine Clonixinate
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C13H11ClN2O2.C6H14N2O2/c1-8-10(14)5-2-6-11(8)16-12-9(13(17)18)4-3-7-15-12;7-4-2-1-3-5(8)6(9)10/h2-7H,1H3,(H,15,16)(H,17,18);5H,1-4,7-8H2,(H,9,10)/t;5-/m.0/s1
Kvantitet 1 g
Formel vikt 408.1564
Procent renhet >95%