Läs mer
L-Lysine L-Malate, TRC
Produktkod. 30476023
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30476023 10 g
1 options
Produktkod. 30476023 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46891010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Lysine L-Malate
CAS 71555-10-7
Molekylformel C6H14N2O2.C4H6O5
Kvantitet 10 g
Synonym L-Lysine (2S)-2-Hydroxybutanedioate, (S)-Hydroxybutanedioic Acid compd. with L-Lysine
InChI formel InChI=1S/C6H14N2O2.C4H6O5/c7-4-2-1-3-5(8)6(9)10;5-2(4(8)9)1-3(6)7/h5H,1-4,7-8H2,(H,9,10);2,5H,1H2,(H,6,7)(H,8,9)/t5-;2-/m00/s1
LEDER NCCCC[C@H](N)C(=O)O.O[C@@H](CC(=O)O)C(=O)O
IUPAC-namn (2S)-2,6-diaminohexanoic acid;(2S)-2-hydroxybutanedioic acid
Molekylvikt (g/mol) 280.27
Formel vikt 280.1271