Läs mer
L-Lyxose, TRC
Produktkod. 30484037
Change view
Kvantitet:
1 g
2.5 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30484037 2.5 g
30467964 5 g
30477414 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30484037 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL5020002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1949-78-6
Molekylformel C5 H10 O5
Molekylvikt (g/mol) 150.13
Synonym L-Lyxose, Lyxose, L- (8CI)
IUPAC-namn (3R,4R,5S)-oxane-2,3,4,5-tetrol
LEDER O[C@H]1COC(O)[C@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material L-Lyxose
Kvantitet 2.5 g
InChI formel InChI=1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3+,4+,5?/m0/s1
Formel vikt 150.0528