Läs mer
L-Mannono-1,4-lactone, TRC
Produktkod. 30478922
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478922 5 g
30471729 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30478922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1660305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22430-23-5
Molekylformel C6 H10 O6
Molekylvikt (g/mol) 178.14
Synonym L-Mannonic acid, γ-lactone, Mannonic acid, γ-lactone, L- (8CI), L-Mannono-1,4-lactone, L-Mannono-γ-lactone
IUPAC-namn (3R,4S,5S)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
LEDER OC[C@H](O)[C@@H]1OC(=O)[C@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material L-Mannonic acid gamma-lactone
Kvantitet 5 g
InChI formel InChI=1S/C6H10O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-5,7-10H,1H2/t2-,3-,4+,5-/m0/s1
Formel vikt 178.0477