Learn More
L-Menthyl 2,2-Dihydroxyacetate, TRC
Product Code. 30478512
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30478512 5 g 5g
1 options
Product Code. 30478512 Supplier Toronto Research Chemicals Supplier No. TRCM2191455G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Menthyl 2,2-Dihydroxyacetate
CAS 111969-64-3
Molecular Formula C12H22O4
Quantity 5 g
Synonym 2,2-Dihydroxyacetic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester, Dihydroxyacetic Acid (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester, [1R-(1alpha,2beta,5alpha)]-Dihydroxyacetic Acid 5-Methyl-2-(1-methylethyl)cyclohexyl Ester, 2,2-Dihydroxyacetic Acid (-)-Menthyl Ester
InChI Formula InChI=1S/C12H22O4/c1-7(2)9-5-4-8(3)6-10(9)16-12(15)11(13)14/h7-11,13-14H,4-6H2,1-3H3/t8-,9+,10-/m1/s1
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)C(O)O
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2,2-dihydroxyacetate
Molecular Weight (g/mol) 230.3
Formula Weight 230.1518