Läs mer
L-Monapterin, TRC
Produktkod. 30134509
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134509 100 mg
30007534 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134509 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM505650100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2277-42-1
Molekylformel C9H11N5O4
Molekylvikt (g/mol) 253.21
Synonym L-threo-1-(2-Amino-4-hydroxy-6-pteridinyl)-1,2,3-propanetriol, [S-(R*,R*)]-2-amino-6-(1,2,3-trihydroxypropyl)-4(1H)pteridinone;' '¡2-Amino-6-[(1S,2S)-1,2,3-trihydroxypropyl]-4(1H)-pteridinone;' '¡2-Aino-6-(L-threo-trihydroxypropyl)-4(3H)-Pteridinone, L-(+)-Monapterin, L-6-(threo-1',2',3'-Trihydroxypropyl)pterin, L-Monapterin, L-threo-Monapterin, L-threo-Neopterin, L-(+)-Monapterin, L-threo-Neopterin;
IUPAC-namn 2-amino-6-((1S,2S)-1,2,3-trihydroxypropyl)pteridin-4(8H)-one
LEDER O=C1C2=NC([C@H](O)[C@@H](O)CO)=CNC2=NC(N)=N1
Specifikationer
Kemiskt namn eller material L-threo-Neopterin
Kvantitet 100 mg
InChI formel InChI=1S/C9H11N5O4/c10-9-13-7-5(8(18)14-9)12-3(1-11-7)6(17)4(16)2-15/h1,4,6,15-17H,2H2,(H3,10,11,13,14,18)/t4-,6-/m0/s1
Formel vikt 253.08