Läs mer
L-NIL, MedChemExpress
Produktkod. 16445756
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16445756 100 mg
16441237 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16445756 Leverantör Medchem Express Leverantörsnummer HY12116
MedChemExpress L-NIL is an inducible NO synthase inhibitor, with an IC50 of 3.3 μM for miNOS.
L-NIL is an inducible NO synthase inhibitor, with an IC50 of 3.3 μM for miNOS.
Specifikationer
Kemiskt namn eller material L-NIL
Fysisk form Solid
CAS 53774-63-3
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C8H17N3O2
För användning med (applikation) COVID-19-immunoregulation
Löslighetsinformation H2O : 50 mg/mL (267.04 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
LEDER N[C@@H](CCCCNC(C)=N)C(O)=O
Molekylvikt (g/mol) 187.24
Formel vikt 187.24
Procent renhet 98.0%