Läs mer
L-Norleucine, TRC
Produktkod. 30010580
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010580 250 mg
30006044 1 g
30003457 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010580 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN712000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 327-57-1
Molekylformel C6 H13 N O2
Molekylvikt (g/mol) 131.17
Synonym L-Norleucine, Norleucine, L- (8CI), (S)-2-Aminohexanoic acid, (S)-Norleucine, (S)-α-Aminohexanoic acid, 2-Aminocaproic acid, 2-Aminohexanoic acid, Caprine, Glycoleucine, Hexanoic acid, 2-amino-, (S)-, L-(+)-Norleucine, L-2-Aminohexanoic acid, NSC 10378, NSC 74430, Norleucine, α-Aminocaproic acid
IUPAC-namn (2S)-2-aminohexanoic acid
LEDER CCCC[C@H](N)C(=O)O
Specifikationer
Kemiskt namn eller material L-Norleucine (Aminocaproic acid)
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/t5-/m0/s1
Kvantitet 250 mg
Formel vikt 131.0946
Procent renhet >95%