Läs mer
L-Orn(Z)-OH, TRC
Produktkod. 30413429
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30413429 1 g
1 options
Produktkod. 30413429 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO6710151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Orn(Z)-OH
CAS 3304-51-6
Molekylformel C13H18N2O4
Kvantitet 1 g
Synonym N5-[(phenylmethoxy)carbonyl]-L-ornithine, (S)-2-Amino-5-(phenylmethoxycarbonylamino)pentanoic Acid, N-delta-Cbz-L-ornithine, N5-(Benzyloxycarbonyl)ornithine, N5-Carboxyornithine N5-Benzyl Ester, Ndelta-Benzyloxycarbonyl-L-ornithine, delta-N-Benzyloxycarbonyl-L-ornithine
InChI formel InChI=1S/C13H18N2O4/c14-11(12(16)17)7-4-8-15-13(18)19-9-10-5-2-1-3-6-10/h1-3,5-6,11H,4,7-9,14H2,(H,15,18)(H,16,17)/t11-/m0/s1
LEDER N[C@@H](CCCNC(=O)OCc1ccccc1)C(=O)O
IUPAC-namn (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid
Molekylvikt (g/mol) 266.29
Formel vikt 266.1267