Läs mer
L-Phenylacetyl Carbinol, TRC
Produktkod. 30134070
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134070 100 mg
30009041 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134070 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP306300100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53439-91-1
Molekylformel C9 H10 O2
Molekylvikt (g/mol) 150.17
Synonym 2-Propanone, 1-hydroxy-1-phenyl-, (S)-, (1S)-1-Hydroxy-1-phenyl-2-propanone, (+)-Phenylacetyl carbinol, (S)-1-Hydroxy-1-phenyl-2-propanone, (S)-1-Hydroxy-1-phenylpropanone, (S)-Phenylacetyl carbinol, L-Phenylacetylcarbinol, (S)-1-Hydroxy-1-phenylpropan-2-one
IUPAC-namn (1S)-1-hydroxy-1-phenylpropan-2-one
LEDER CC(=O)[C@@H](O)c1ccccc1
Specifikationer
Kemiskt namn eller material L-Phenylacetyl Carbinol
Kvantitet 100 mg
InChI formel InChI=1S/C9H10O2/c1-7(10)9(11)8-5-3-2-4-6-8/h2-6,9,11H,1H3/t9-/m1/s1
Formel vikt 150.0681