Learn More
Thermo Scientific Chemicals L-Phenylalanine ethyl ester hydrochloride, 99%
Catalog Number p-181800
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
3182-93-2
C11H16ClNO2
229.70
MFCD00012507
FPFQPLFYTKMCHN-PPHPATTJSA-N
l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
165085
ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
3182-93-2
C11H16ClNO2
229.70
MFCD00012507
FPFQPLFYTKMCHN-PPHPATTJSA-N
l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
165085
ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 154°C to 157°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CH2CH(NH2)CO2C2H5·HCl
Quantity 10 g
Beilstein 14, 497
Specific Rotation Condition +33.70° (21°C c=2,C2H5OH)
Specific Rotation + 33.7
Formula Weight 229.71
Percent Purity 99%
Physical Form Fine Needle-like Crystalline Solid
Chemical Name or Material L-Phenylalanine ethyl ester hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • L-Phenylalanine ethyl ester hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.