Läs mer
L-Pyroglutamic Acid Beta-Naphthylamide, TRC
Produktkod. 30423352
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423352 100 mg
30410810 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423352 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP997500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22155-91-5
Molekylformel C15 H14 N2 O2
Molekylvikt (g/mol) 254.28
Synonym L-Pyroglutamic Acid -Ss-Naphthylamide
IUPAC-namn (2S)-N-naphthalen-2-yl-5-oxopyrrolidine-2-carboxamide
LEDER O=C(Nc1ccc2ccccc2c1)[C@@H]3CCC(=O)N3
Specifikationer
Kemiskt namn eller material L-Pyroglutamic Acid beta-Naphthylamide
Kvantitet 100 mg
InChI formel InChI=1S/C15H14N2O2/c18-14-8-7-13(17-14)15(19)16-12-6-5-10-3-1-2-4-11(10)9-12/h1-6,9,13H,7-8H2,(H,16,19)(H,17,18)/t13-/m0/s1
Formel vikt 254.1055