missing translation for 'onlineSavingsMsg'
Läs mer

L-Ribulose, TRC

Produktkod. 30415539
Change view
Click to view available options
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30415539 10 mg
30413680 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30415539 Supplier Toronto Research Chemicals Supplier No. TRCR41800310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2042-27-5
Molekylformel C5 H10 O5
Molekylvikt (g/mol) 150.13
Synonym L-erythro-Pentulose
IUPAC-namn (3S,4S)-1,3,4,5-tetrahydroxypentan-2-one
LEDER OC[C@H](O)[C@H](O)C(=O)CO

Specifications

Kemiskt namn eller material L-Ribulose
Kvantitet 10 mg
InChI formel InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h3,5-8,10H,1-2H2/t3-,5-/m0/s1
Formel vikt 150.0528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.