Läs mer
L-Sepiapterin, TRC
Produktkod. 30132922
Change view
Kvantitet:
0.5 mg
1 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30132922 0.5 mg 0.5mg
30135268 5 mg -
30005611 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2589130.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17094-01-8
Molekylformel C9H11N5O3
Molekylvikt (g/mol) 237.22
Synonym Sepiapterine, 2-Amino-7,8-dihydro-6-[(2S)-2-hydroxy-1-oxopropyl]-4(3H)-pteridinone
IUPAC-namn (2S)-1-(2-amino-4-hydroxy-7,8-dihydropteridin-6-yl)-2-hydroxypropan-1-one
LEDER O=C(N=C(N)N1)C2=C1NCC(C([C@H](O)C)=O)=N2
Specifikationer
Kemiskt namn eller material L-Sepiapterin
Kvantitet 0.5 mg
InChI formel InChI=1S/C9H11N5O3/c1-3(15)6(16)4-2-11-7-5(12-4)8(17)14-9(10)13-7/h3,15H,2H2,1H3,(H4,10,11,13,14,17)/t3-/m0/s1
Formel vikt 237.0862