missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Tartaric acid diammonium salt, 99%, Thermo Scientific Chemicals

Catalog Number p-194817
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
3164-29-2
C4H12N2O6
184.15
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
This item is not returnable. View return policy
3164-29-2
C4H12N2O6
184.15
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Density 1.6000g/mL
pH 6.0 to 7.0 (5% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Linear Formula [-CH(OH)CO2NH4]2
Quantity 5 g
Merck Index 09, 591
Specific Rotation Condition + 32.40 (20.00°C c=1.84,H2O)
Specific Rotation +31.4° to +33.4° (20°C, 589nm) (c=1.84, H2O)
IUPAC Name diazanium;(2R,3R)-2,3-dihydroxybutanedioate
Formula Weight 184.15
Percent Purity ≥98.5%
Insoluble Matter 0.005% max.
Physical Form Crystals
Chemical Name or Material L(+)-Tartaric acid diammonium salt, 0.99
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'