Läs mer
L-(+)-threo-Chloramphenicol, TRC
Produktkod. 30358125
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358125 100 mg
30350000 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358125 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC325025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 134-90-7
Molekylformel C11 H12 Cl2 N2 O5
Molekylvikt (g/mol) 323.13
Synonym 2,2-Dichloro-N-[(1S,2S)-2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]acetamide, Acetamide, 2,2-dichloro-N-[2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]-, [S-(R*,R*)]-, Acetamide, 2,2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-nitrophenethyl]-, L-threo- (6CI), Acetamide, 2,2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-nitrophenethyl]-, L-threo-(+)- (8CI), (+)-Chloramphenicol, (1S,2S)-2-(2,2-Dichloroacetamido)-1-(4-nitrophenyl)-1,3-propanediol, Dextramycin, Dextromycetin, L-(+)-threo-Chloramphenicol, L-threo-(1S,2S)-1-p-Nitrophenyl-2-dichloroacetamido-1,3-propanediol, L-threo-Chloramphenicol, L-threo-Chloroamphenicol, L-threo-N-Dichloracetyl-1-p-nitrophenyl-2-amino-1,3-propanediol, L-threo-N-Dichloroacetyl-1-p-nitrophenyl-2-amino-1,3-propanediol
IUPAC-namn 2,2-dichloro-N-[(1S,2S)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
LEDER OC[C@H](NC(=O)C(Cl)Cl)[C@@H](O)c1ccc(cc1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material L-(+)-threo-Chloramphenicol
Kvantitet 100 mg
InChI formel InChI=1S/C11H12Cl2N2O5/c12-10(13)11(18)14-8(5-16)9(17)6-1-3-7(4-2-6)15(19)20/h1-4,8-10,16-17H,5H2,(H,14,18)/t8-,9-/m0/s1
Formel vikt 322.0123