Läs mer
L-threo-Droxidopa, TRC
Produktkod. 30372906
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30372906 250 mg
1 options
Produktkod. 30372906 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD689960250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-threo-Droxidopa
CAS 23651-95-8
Molekylformel C9 H11 N O5
Kvantitet 250 mg
Synonym L-Tyrosine, β,3-dihydroxy-, (βR)-, L-Tyrosine, β,3-dihydroxy-, threo-, Serine, 3-(3,4-dihydroxyphenyl)-, L-threo- (8CI), (βR)-β,3-Dihydroxy-L-tyrosine, (-)-threo-3,4-Dihydroxyphenylserine, (2S,3R)-3-(3,4-Dihydroxyphenyl)-2-amino-3-hydroxypropanoic acid, DOPS, Droxidopa, L-DOPS, L-Threodops, L-threo-3,4-Dihydroxyphenylserine, L-threo-3-(3,4-Dihydroxyphenyl)serine, L-threo-DOPS, L-threo-β-(3,4-Dihydroxyphenyl)serine, Northera, SM 5688, threo-3-(3,4-Dihydroxyphenyl)-L-serine, threo-Dopaserine
InChI formel InChI=1S/C9H11NO5/c10-7(9(14)15)8(13)4-1-2-5(11)6(12)3-4/h1-3,7-8,11-13H,10H2,(H,14,15)/t7-,8+/m0/s1
LEDER N[C@@H]([C@H](O)c1ccc(O)c(O)c1)C(=O)O
IUPAC-namn (2S,3R)-2-amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropanoic acid
Molekylvikt (g/mol) 213.19
Formel vikt 213.0637