Läs mer
L-threo-Hex-2-enaric acid 1,4-Lactone, TRC
Produktkod. 30008045
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008045 50 mg
17978214 25 mg
17909064 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008045 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH29483050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66757-69-5
Molekylformel C6 H6 O7
Molekylvikt (g/mol) 190.11
Synonym L-threo-Hex-2-enaric acid, 1,4-lactone (9CI), (R)-[(2R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl]hydroxyacetic Acid
IUPAC-namn (2R)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyacetic acid
LEDER O[C@H]([C@H]1OC(=O)C(=C1O)O)C(=O)O
Specifikationer
Kemiskt namn eller material L-threo-Hex-2-enaric acid 1,4-Lactone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C6H6O7/c7-1-2(8)6(12)13-4(1)3(9)5(10)11/h3-4,7-9H,(H,10,11)/t3-,4+/m1/s1
Formel vikt 190.0114
Procent renhet >95%