Läs mer
L-Threose (~0.2 M in Water), TRC
Produktkod. 30455599
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455599 100 mg
30454528 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30455599 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT405515100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 95-44-3
Molekylformel C4H8O4
Molekylvikt (g/mol) 120.1
Synonym (2R,3S)-2,3,4-Trihydroxy-butanal, [R-(R*,S*)]-2,3,4-Trihydroxy-butanal, L-Threofuranose
IUPAC-namn (2R,3S)-2,3,4-trihydroxybutanal
LEDER OC[C@H](O)[C@@H](O)C=O
Specifikationer
Kemiskt namn eller material L-Threose
Kvantitet 100 mg
InChI formel InChI=1S/C4H8O4/c5-1-3(7)4(8)2-6/h1,3-4,6-8H,2H2/t3-,4-/m0/s1
Formel vikt 120.0423
Koncentration eller sammansättning (efter analyt eller komponenter) 0.2 M