Läs mer
L-Thyronine, 95%, TRC
Produktkod. 17994685
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994685 250 mg
17960755 100 mg
17972855 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17994685 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT922700250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1596-67-4
Molekylformel C15 H15 N O4
Molekylvikt (g/mol) 273.28
Synonym L-Thyronine, 95%, O-(4-Hydroxyphenyl)-L-tyrosine, Desiodothyroxine, β-(p-Hydroxyphenoxy)phenylalanine
IUPAC-namn (2S)-2-amino-3-[4-(4-hydroxyphenoxy)phenyl]propanoic acid
LEDER N[C@@H](Cc1ccc(Oc2ccc(O)cc2)cc1)C(=O)O
Specifikationer
Kemiskt namn eller material L-Thyronine
InChI formel InChI=1S/C15H15NO4/c16-14(15(18)19)9-10-1-5-12(6-2-10)20-13-7-3-11(17)4-8-13/h1-8,14,17H,9,16H2,(H,18,19)/t14-/m0/s1
Kvantitet 250 mg
Formel vikt 273.1001