Läs mer
L-Tryptophan-13C11-15N2, TRC
Produktkod. 30011485
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011485 2.5 mg 2.5mg
30004431 10 mg -
30004758 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2024062.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 202406-50-6
Molekylformel 13C11H1215N2O2
Molekylvikt (g/mol) 217.13
Synonym L-Tryptophan-carboxy,α,β,2,3,3a,4,5,6,7,7a-13C11-Nα,N1-15N2, L-Tryptophan-13C11-15N2 (9CI)
LEDER [15NH2][13C@H]([13C](O)=O)[13CH2][13C]1=[13CH][15NH][13C]2=[13C]1[13CH]=[13CH][13CH]=[13CH]2
Specifikationer
Kemiskt namn eller material L-Tryptophan-13C11-15N2
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1
Formel vikt 217.1209
Procent renhet >95%