Läs mer
L-Tryptophan-15N2, TRC
Produktkod. 30009619
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009619 25 mg
30010955 50 mg
30005136 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT20463425MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 204634-20-8
Molekylformel C11H1215N2O2
Molekylvikt (g/mol) 206.21
Synonym L-Tryptophan-15N2, L-[15N2]-Tryptophan, (S)-Tryptophan-15N2, (S)-α-(Amino-15N)-3-(indole-15N)propionic acid, (S)-α-Aminoindole-3-propanoic acid 15N2
IUPAC-namn (2S)-2-(^{15}N)azanyl-3-(1H-indol-3-yl)propanoic acid
LEDER [15NH2][C@H](C(O)=O)CC1=C[15N]([H])C2=C1C=CC=C2
Specifikationer
Kemiskt namn eller material L-Tryptophan-15N2
Kvantitet 25 mg
InChI formel InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1/i12+1,13+1
Formel vikt 206.0839