Läs mer
L-Tryptophyl-L-arginyl-L-glutaminyl-L-alanyl-L-alanyl-L-phenylalanyl-L-valyl-L-Alpha-aspartyl-L-seryl-L-tyrosine TFA Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Tryptophyl-L-arginyl-L-glutaminyl-L-alanyl-L-alanyl-L-phenylalanyl-L-valyl-L-alpha-aspartyl-L-seryl-L-tyrosine TFA Salt |
| CAS | 955091-53-9 |
| Molekylformel | C58H79N15O16 .XC2HF3O2 |
| Kvantitet | 1 mg |
| Synonym | 10Panx TFA Salt |
| InChI formel | InChI=1S/C58H79N15O16/c1-29(2)47(56(87)70-42(26-46(77)78)53(84)72-44(28-74)55(86)71-43(57(88)89)24-33-16-18-35(75)19-17-33)73-54(85)41(23-32-11-6-5-7-12-32)69-49(80)31(4)65-48(79)30(3)66-51(82)40(20-21-45(60)76)68-52(83)39(15-10-22-63-58(61)62)67-50(81)37(59)25-34-27-64-38-14-9-8-13-36(34)38/h5-9,11-14,16-19,27,29-31,37,39-44,47,64,74-75H,10,15,20-26,28,59H2,1-4H3,(H2,60,76)(H,65,79)(H,66,82)(H,67,81)(H,68,83)(H,69,80)(H,70,87)(H,71,86)(H,72,84)(H,73,85)(H,77,78)(H,88,89)(H4,61,62,63) |
| LEDER | CC(C)C(NC(=O)C(Cc1ccccc1)NC(=O)C(C)NC(=O)C(C)NC(=O)C(CCC(=O)N)NC(=O)C(CCCN=C(N)N)NC(=O)C(N)Cc2c[nH]c3ccccc23)C(=O)NC(CC(=O)O)C(=O)NC(CO)C(=O)NC(Cc4ccc(O)cc4)C(=O)O |
| IUPAC-namn | 3-[[2-[[2-[2-[2-[[5-amino-2-[[2-[[2-amino-3-(1H-indol-3-yl)propanoyl]amino]-5-(diaminomethylideneamino)pentanoyl]amino]-5-oxopentanoyl]amino]propanoylamino]propanoylamino]-3-phenylpropanoyl]amino]-3-methylbutanoyl]amino]-4-[[1-[[1-carboxy-2-(4-hydroxyphenyl)ethyl]amino]-3-hydroxy-1-oxopropan-2-yl]amino]-4-oxobutanoic acid |
| Molekylvikt (g/mol) | 1242.3 |
| Formel vikt | 1241.5829 |