Läs mer
L-Tyrosine-13C, TRC
Produktkod. 30455144
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455144 50 mg
30450346 5 mg
30452437 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455144 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT96639850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81201-89-0
Molekylformel C813CH11NO3
Molekylvikt (g/mol) 182.18
Synonym L-Tyrosine-carboxy-13C (9CI, ACI), L-Tyrosine-1-13C, L-Tyrosine-13C, (2S)-2-Amino-3-(4-hydroxyphenyl)propanoic-1-13C acid, 4-Hydroxyphenyl-L-alanine-1-13C, β-(p-Hydroxyphenyl-d4)alanine-carboxy-13C, L-4-Hydroxyphenylalanine-carboxy-13C, L-Tyrosine-13C (carboxy-13C)
LEDER OC1=CC=C(C[C@H](N)[13C](O)=O)C=C1
Specifikationer
Kemiskt namn eller material L-Tyrosine-13C
InChI formel InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1/i9+1
Kvantitet 50 mg
Formel vikt 182.0772