Läs mer
L-Tyrosine-13C6, TRC
Produktkod. 30135280
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135280 50 mg
30011316 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135280 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT89997750MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 201595-63-3
Molekylformel C313C6H11NO3
Molekylvikt (g/mol) 187.14
Synonym (-)-α-Amino-p-hydroxyhydrocinnamic Acid-13C6, Therigon-13C6, p-Tyrosine-13C6, (2S)-2-Amino-3-(4-hydroxyphenyl)propanoic Acid-13C6, (S)-2-Amino-3-(4-hydroxyphenyl)propanoic Acid-13C6, (S)-Tyrosine-13C6, (S)-α-Amino-4-hydroxybenzenepropanoic Acid-13C6, L-(-)-Tyrosine-13C6, NSC 82624-13C6, NSC 9973-13C6;
LEDER N[C@H](C(O)=O)C[13C]1=[13C]([H])[13C]([H])=[13C](O)[13C]([H])=[13C]1[H]
Specifikationer
Kemiskt namn eller material L-Tyrosine-13C6
Kvantitet 50 mg
InChI formel InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1/i1+1,2+1,3+1,4+1,6+1,7+1
Formel vikt 187.094