Läs mer
L-tyrosin bensylester 4-toluensulfonat, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Tyrosine Benzyl Ester 4-Toluenesulfonate |
| CAS | 53587-11-4 |
| Molekylformel | C16H17NO3 . C7H8O3S |
| Kvantitet | 50 mg |
| Synonym | L-Tyrosine','phenylmethyl ester','4-methylbenzenesulfonate (1:1), L-Tyrosine','phenylmethyl ester','4-methylbenzenesulfonate (salt) (9CI), (S)-Tyrosine benzyl ester tosylate, Benzyl L-tyrosinate tosylate, L-Tyrosine benzyl ester 4-methylbenzenesulfonate, L-Tyrosine benzyl ester p-toluenesulfonate, Tyrosine benzyl ester tosylate, Tyrosine benzyl ester tosylate (salt) |
| InChI formel | InChI=1S/C16H17NO3.C7H8O3S/c17-15(10-12-6-8-14(18)9-7-12)16(19)20-11-13-4-2-1-3-5-13;1-6-2-4-7(5-3-6)11(8,9)10/h1-9,15,18H,10-11,17H2;2-5H,1H3,(H,8,9,10)/t15-;/m0./s1 |
| LEDER | Cc1ccc(cc1)S(=O)(=O)O.N[C@@H](Cc2ccc(O)cc2)C(=O)OCc3ccccc3 |
| IUPAC-namn | bensyl (2S)-2-amino-3-(4-hydroxyfenyl)propanoat; 4-metylbensensulfonsyra |
| Molekylvikt (g/mol) | 443.51 |
| Formel vikt | 443.1403 |