missing translation for 'onlineSavingsMsg'
Läs mer

L-Tyrosine Disodium Salt Dihydrate, TRC

Produktkod. 30009385
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009385 100 mg
30005309 250 mg
30012383 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30009385 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL218310100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 122666-87-9
Molekylformel C9H9NO3Na2 . 2H2O
Molekylvikt (g/mol) 261.18
Synonym L-Tyrosine, sodium salt, hydrate (1:2:2) (ACI), L-Tyrosine, disodium salt, dihydrate (9CI), L-Tyrosine disodium salt dihydrate, (2S)-2-Amino-3-(4-hydroxyphenyl)propanoic acid disodium salt dihydrate
IUPAC-namn disodium;(2S)-2-amino-3-(4-oxidophenyl)propanoate;dihydrate
LEDER O.O.[Na+].[Na+].N[C@@H](Cc1ccc([O-])cc1)C(=O)[O-]

Specifikationer

Kemiskt namn eller material L-Tyrosine Disodium Salt Dihydrate
InChI formel InChI=1S/C9H11NO3.2Na.2H2O/c10-8(9(12)13)5-6-1-3-7(11)4-2-6;;;;/h1-4,8,11H,5,10H2,(H,12,13);;;2*1H2/q;2*+1;;/p-2/t8-;;;;/m0..../s1
Kvantitet 100 mg
Formel vikt 261.0589
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.