Läs mer
L-Valine-13C, TRC
Produktkod. 30012454
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012454 50 mg
30004640 10 mg
30011482 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV94341050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81201-85-6
Molekylformel C413CH11NO2
Molekylvikt (g/mol) 118.14
Synonym L-Valine-1-13C (9CI, ACI), L-[1-13C]Valine, [1-13C]-L-Valine, 2-Amino-3-methylbutanoic-1-13C acid, L-Valine-13C
LEDER CC(C)[C@@H]([13C](=O)O)N
Specifikationer
Kemiskt namn eller material L-Valine-13C
Kvantitet 50 mg
InChI formel InChI=1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i5+1
Formel vikt 118.0823