Läs mer
L-Valine Benzyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Valine Benzyl Ester |
| CAS | 21760-98-5 |
| Molekylformel | C12 H17 N O2 |
| Kvantitet | 1 g |
| Synonym | L-Valine','phenylmethyl ester, Valine','benzyl ester','L- (8CI), (S)-Benzyl 2-amino-3-methylbutanoate, Benzyl L-valinate, Benzyl L-valinate, L-Valine benzyl ester, Valine benzyl ester, (2S)-2-Amino-3-methylbutanoic acid phenylmethyl ester |
| InChI formel | InChI=1S/C12H17NO2/c1-9(2)11(13)12(14)15-8-10-6-4-3-5-7-10/h3-7,9,11H,8,13H2,1-2H3/t11-/m0/s1 |
| LEDER | CC(C)[C@H](N)C(=O)OCc1ccccc1 |
| IUPAC-namn | benzyl (2S)-2-amino-3-methylbutanoate |
| Molekylvikt (g/mol) | 207.27 |
| Formel vikt | 207.1259 |