Läs mer
L-Valyl-L-histidyl-L-leucyl-L-threonyl-L-prolyl-L-glutamic Acid Trifluoroacetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Valyl-L-histidyl-L-leucyl-L-threonyl-L-prolyl-L-glutamic Acid Trifluoroacetate |
| CAS | 93913-39-4 |
| Molekylformel | C31H50N8O10 . xC2HF3O2 |
| Kvantitet | 100 mg |
| Synonym | N-[1-[N-[N-(N-Valylhistidyl)leucyl]threonyl]prolyl]-glutamic Acid Trifluoroacetate, 1: PN: WO2010067611 SEQID: 1 Claimed Protein, 1: PN: WO2010067612 SEQID: 1 Unclaimed Protein, 9: PN: WO2011072833 SEQID: 3 Claimed Protein |
| InChI formel | InChI=1S/C31H50N8O10/c1-15(2)11-20(36-26(43)21(12-18-13-33-14-34-18)37-29(46)24(32)16(3)4)27(44)38-25(17(5)40)30(47)39-10-6-7-22(39)28(45)35-19(31(48)49)8-9-23(41)42/h13-17,19-22,24-25,40H,6-12,32H2,1-5H3,(H,33,34)(H,35,45)(H,36,43)(H,37,46)(H,38,44)(H,41,42)(H,48,49) |
| LEDER | CC(C)C[C@H](NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@@H](N)C(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N2CCC[C@H]2C(=O)NC(CCC(=O)O)C(=O)O.OC(=O)C(F)(F)F |
| Molekylvikt (g/mol) | 694.8 |