Läs mer
L-xylo-2-hexulosonsyra metylester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl L-Sorbosonate |
| CAS | 3031-98-9 |
| Molekylformel | C7 H12 O7 |
| Kvantitet | 250 mg |
| Synonym | L-xylo-2-Hexulosonic acid','methyl ester, Gulonic acid','2-keto-','methyl ester','L- (6CI), L-xylo-Hexulosonic acid','methyl ester (7CI,8CI), 2-keto-L-Gulonic acid methyl ester, Methyl 2-keto-L-gulonate, Methyl L-xylo-hexulosonate, NSC 163924, Methyl L-Sorbosonate, Methyl L-xylo-Hex-2-ulosonate, Ascorbic Acid Imp. D (EP) |
| InChI formel | InChI=1S/C7H12O7/c1-14-7(13)6(12)5(11)4(10)3(9)2-8/h3-5,8-11H,2H2,1H3/t3-,4+,5-/m0/s1 |
| LEDER | COC(=O)C(=O)[C@@H](O)[C@H](O)[C@@H](O)CO |
| IUPAC-namn | metyl (3S,4R,5S)-3,4,5,6-tetrahydroxy-2-oxohexanoat |
| Molekylvikt (g/mol) | 208.166 |
| Formel vikt | 208.0583 |