Learn More
Hettich Lab Technology™ Swing-Out Rotor with Carriers
Hettich Lab Technology™ Swing-Out Rotor with Carriers is suitable for performing rate-zonal and isopycnic centrifugations
Brand:  Hettich Lab Technology™ 1619
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10597861
  • 8840.00 SEK / Each
Estimated Shipment: 29-05-2024
to see stock.
Description
Description
  • Complete with carriers
  • Convenient and easy to use
  • Built to withstand long-term, repeated use
Specifications
Specifications
Swing-out Rotor
2,701 x g
151 mm
ROTOFIX 32 A and Universal 320/320 R Centrifuges
Carriers
15 x 15 mL
4,000 rpm
120 mm
90°
6