Läs mer
Cyclin D2 Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry experiments.
Brand: Epredia MS-213-P1
Beskrivning
Cyclin D2 is a G1 cyclin required for G1-phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk4 and/or cdk6.Host Species: Mouse
Clone: DCS-3.1
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Purified human recombinant full-length cyclin D2 protein
Molecular Weight: 34 kDa
Positive Control: U-2-OS, C82, RD, Bristol-8 or LoVo36 cells. Note that T47D, ZR75, SKBR3, MCF-7, BT-20, MDA468, HBL100, BT5549, HeLa, Saos2 and HT29 cells are negative for cyclin D2.
Cellular Localization: Nuclear
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Biological Blockade
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifikationer
| Cyclin D2 Ab-2 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Cell Cycle | |
| Human, Mouse, Rat | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| DCS-3.1 | |
| Unconjugated | |
| Purified human recombinant full length cyclin D2 protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.