missing translation for 'onlineSavingsMsg'
Läs mer

Epredia™ Lab Vision™ Milk Fat Globule 1 Human (HMFG1) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Produktkod. 12631597 Handla allt Epredia Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 μL
Form:
Concentrated
Förpackningsstorlek:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Form unitSize
12631597 500 μL Concentrated 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 12631597 Leverantör Epredia™ Leverantörsnummer MS512P1

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Epredia™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Human milk fat globule membrane protein (HMFG1) is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as on the outer surface of the human milk fat globule. HMFG1 is considered as a differentiation marker. It is useful as specific breast epithelial marker in normal and neoplastic mammary development.

Host Species: Mouse

Clone: EDM45

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Delipidated human milk fat globule membrane preparation

Molecular Weight: 40-45kDa

Positive Control: SKBR-3 cells; Breast, colon, or lung tissues

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Milk Fat Globule 1, Human (HMFG1) Ab-1
Användningsområden Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassificering Monoclonal
Klona EDM45
Koncentration 200μg/mL
Konjugera Unconjugated
Värd art Mouse
Immunogen Delipidated human milk fat globule membrane preparation
Kvantitet 500 μL
Regulatorisk status RUO
Forskningsdisciplin Cancer and Tumor Biology
Primär eller sekundär Primary
Målarter Human
Form Concentrated
Isotyp IgG2a κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.