Läs mer
Labconco™ Clear Drying Chambers for FreeZone™ Freeze-Dry Systems
For visible bulk drying
Beskrivning
For visible bulk drying
- Durable and corrosion-resistant
- Chambers without valves include clear acrylic top and bottom lids, gaskets, vacuum break valve, and electrical pass-through for transformer cords
- Chambers with valves include aluminum lid with eight valves that accommodate both 1/2 in. and 3/4 in. adapters for connecting flasks
- Use with or without product shelves (shelves available separately, visit www.fishersci.com)