Learn More
Labconco™ FilterMate™ Exhauster Filter Cells
Replacement filters for use with Labconco FilterMate Portable Exhausters.
Brand:  Labconco™ 3923400
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10646403
  • 3580.00 SEK / Each
Estimated Shipment: 13-06-2024
to see stock.
Description
Description
Organic Carbon Filter, Type AC, contains 12 pounds of granular activated carbon media suitable for organic solvents including acetone, ethyl acetate, toluene and xylene. For use with FilterMate Portable Exhausters.
Specifications
Specifications
Carbon
2.1 in.
7.7 Kg
18 in.
5.3 cm
45.7 cm
45.7 cm
Organic solvents including acetone, ethyl acetate, toluene and xylene
FilterMate Portable Exhauster
17 lb.
10 to 20 in.
18 in.
10 to 20 x 18 x 2.1 in. (25.4 to 50.8 x 45.7 x 5.3 cm)
25.4 to 50.8 cm
Filter