Läs mer
Labconco™ Freeze Dry Manifold
Handla allt Labconco Corporation Produkter
Inkluderar:
20-ports grenrör
Förpackningsstorlek:
Varje
Beskrivning
Drying accessories for FreeZone Freeze Dryers.
- Drying accessories for FreeZone Freeze Dryers.
- Made of stainless steel.
- Include neoprene valves with plastic knobs.
4-Port Manifold:
- 4 valves accommodate both 0.5 or 0.75 in. adapters.
- For use with FreeZone™ 1, 2.5, and 4.5 L Freeze-Dry Systems.
- Dimensions (L x W x H): 8.75 x 8.75 x 9 in. (22.1 x 22.1 x 22.9 cm)
- Attachment Port Lid Accessory required for connection to 4.5 L System (sold separately)
20-Port Manifold:
- 20 neoprene valves that accommodate both 0.5 or 0.75 in. adapters.
- For use with all FreeZone Freeze-Dry Systems.
- Dimensions (L x W x H): 5 x 19 x 9 in. (13 x 48 x 23 cm)
- Attachment Port Lid Accessory required for connection to 4.5 L System (sold separately).
10-Port Manifold with Support Shelf:
- 10 neoprene valves accommodate both 0.5 or 0.75 in. adapters and adjustable support shelf.
- For use with FreeZone 6, 12, and 18 L Freeze-Dry Systems.
- Dimensions (L x W x H): 6.62 x 26 x 10 in. (17 x 66 x 25 cm)
20-Port Manifold with Support Shelves:
- 20 neoprene valves accommodate both 0.5 or 0.75 in. adapters and two adjustable support shelves.
- For use with FreeZone 6, 12, and 18 L Freeze Dry Systems.
- Dimensions (L x W x H): 9.5 x 26 x 10 in. (24 x 66 x 25 cm)
Specifikationer
Specifikationer
| Typ | Freeze Dry Manifold |
| Kapacitet (metrisk) | 1, 2.5, 4.5 L |
| Antal hyllor | 0 |
| Material | Rostfritt stål |
| Antal hamnar | 20 |
| För användning med (utrustning) | 1 L, 2.5 L, 4.5 L, 6 L, 12 L and 18 L FreeZone™ Freeze Dry Systems |
| Inkluderar | 20-ports grenrör |
| Bredd (engelska) | 19 tum |
| Höjd (engelska) | 11 tum |
| Höjd (metrisk) | 27.94 cm |
| Visa mer |