Learn More
Labconco™ General Glove Box: Mechanical/Electrical Straight Pass–Through Kits
Accessories for Labconco General Glove Box
690.00 SEK - 2640.00 SEK
Specifications
For Use With (Equipment) | Precise™ Glove Box, XPert™ Weigh Box |
---|---|
Product Type | Mechanical or Electrical Straight Pass-through Kit |
Product Code | Brand | Product Type | Weight (English) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Weight (English) | Includes | Price | Quantity & Availability | |||
13428469
|
Labconco™
5240700 |
Mechanical or Electrical Straight Pass-through Kit | 0.5 lb. | 3 Pass through Ports with 0.23/0.39, 0.23/0.53 and 0.45/0.70 I.D. Ranges |
690.00 SEK
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
13458469
|
Labconco™
5240800 |
Mechanical or Electrical Straight Pass-through Kit | 1 lb. | – |
2640.00 SEK
Each |
Estimated Shipment: 08-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
For use with Labconco General Glove Box
Specifications
Precise™ Glove Box, XPert™ Weigh Box | |
Mechanical or Electrical Straight Pass-through Kit |