Learn More
Labconco™ Single Electrical Receptacle Kits
Mount on the left or right side of any 3, 4, 5, 6, 7 or 8 ft. Protector Premier, XStream, XL, PVC Perchloric Acid or Stainless Steel Laboratory Hood
Brand: Labconco™ 9412700
Additional Details : Weight : 1.80000kg
Includes: Electrical receptacle, wiring, junction box and receptacle cover plate
Description
- 4 lb. (1.8kg)
- 12L × 9W × 2 in. H (30.5 × 22.9 × 5.1cm)
- Available in choice of different outlets that serve different electrical requirements
- International GFCI Switch also available separately
Specifications
Protector™ Premier™, XStream™, XL™ fume hood | |
4 lb. | |
9 in. | |
Electrical Receptacle, Wiring, Junction Box, Receptacle Cover Plate | |
Single Electrical Receptacle Kit | |
230 V, 50 Hz, 16 A (Schuko Plug) | |
50 Hz |
2 in. | |
1.8 Kg | |
5.1 cm | |
22.9 cm | |
230 V | |
Mounting on Left or Right Side of Fume Hood | |
Schuko Plug |