Learn More
Labconco™ SteamScrubber™ Culture Tube Inserts
Culture Tube Inserts for use with Labconco FlaskScrubber and SteamScrubber Glassware Washers.
Brand:  Labconco™ 4401401
Additional Details : Weight : 0.68000kg
 View more versions of this product
Product Code. 15189655
  • 795.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
Type 304 stainless steel top is for use with Culture Tube Inserts 4588600, 4402001 and 4402101. One Retainer Top is included with Culture Tube Insert 4588600.
Specifications
Specifications
21.33cm
Culture tube insert of FlaskScrubber™, SteamScrubber™ glassware washer
0.7cm
0.7kg
Each
Type 304 Stainless Steel
Retainer Top
46.9cm