Learn More
Labconco™ SteamScrubber™ Labware Inserts
Hold a variety of wider-mouth flasks, beakers and other labware in inverted position
Brand:  Labconco™ 4401801
Additional Details : Weight : 1.14000kg
 View more versions of this product
Product Code. 15179655
  • 2355.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Specifications
Specifications
22.35cm
Hold a variety of wider mouth flasks, beakers and other labware in inverted position
Type 304 Stainless Steel, Plastic
32.0
1.1kg
Each
FlaskScrubber™, SteamScrubber™ glassware washer
15.24cm
Labware Insert
49cm