Learn More
Labconco™ Storage Shelf Kit for Precise™ Glove Boxes
Accessories for use with Precise Glove Boxes
Brand: Labconco™ 5235000
Additional Details : Weight : 1.00000kg
Description
- Rear interior
- Provides additional usable surface area for small items inside glove box
- Adds 240 sq. in. of usable space
- Consists of 4 SS shelves, 2 epoxy-coated steel vertical uprights, SS slotted back
Specifications
Precise™ Basic™ Glove Box | |
2.3 Kg | |
Storage Shelf Kit |
5 lb. | |
4 Stainless Steel Shelves, 2 Epoxy Coated Steel Vertical Uprights, Stainless Steel Slotted Back |