missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Washer Racks

For use with Labconco FlaskScrubber™ and SteamScrubber™ Washers

Brand:  Labconco™ 4595900

Additional Details : Weight : 7.27000kg

 View more versions of this product

Product Code. 15169685

  • 8125.00 SEK / Each


Estimated Shipment
23-11-2020
Add to basket

Description

Description

Type 304 stainless-steel construction

Upper Spindle Rack

  • Provides direct injection cleaning and forced air drying through the spindles
  • Accommodates glassware up to 8.9 in.H (22.6cm)
  • With side release latches and side rails for tank side wall installation
  • Includes 30 removable spindles, 30 adjustable-height clips, 10 small spindles, 15 plastic glassware folders, tool, 30 nylon plugs, and wash arm
  • Spindles may be removed and plugged to accommodate various accessory inserts (sold separately)
Lower Spindle Rack
Provides direct injection cleaning through the spindles
Accommodates glassware up to 18.7 in.H (47.5cm) without upper rack in place and 8.3 - 10.6 in.H (21.1 x 26.9cm) with upper rack in place, depending on rack position
May be used in place of the Lower Standard Rack in any SteamScrubber
Includes 36 removable spindles, 36 adjustable height clips, 10 small spindles, tool to remove spindles and 36 nylon plugs
Spindles may be removed and plugged to accommodate accessory inserts (sold separately)
Upper and Lower Standard Racks
  • Supplied with SteamScrubber washers
  • Can be used with FlaskScrubber and FlaskScrubber Vantage™ Series washers
  • With release latches and wash arm
  • Accessory basket inserts (sold separately) can be added to suit glassware washing needs

Specifications

Specifications

Type 304 Stainless Steel
Each
Side release latches and wash arm
Upper Standard Rack
51.3cm
53cm
FlaskScrubber™, FlaskScrubber™ Vantage series glassware washer
15.2cm
7.25kg
Videos
Documents