Läs mer
Labetalol Hydrochloride, TRC
Produktkod. 17932875
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17932875 50 mg
17954385 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17932875 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL09649550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32780-64-6
Molekylformel C19 H24 N2 O3 . Cl H
Molekylvikt (g/mol) 364.87
Synonym Benzamide, 2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, hydrochloride (1:1), Benzamide, 2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, monohydrochloride (9CI), Salicylamide, 5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, monohydrochloride (8CI), AH 5158 hydrochloride, AH 5158A, Amipress, Ipolab, Labelol, Labetalol hydrochloride, Labracol, NSC 290312, Normodyne, Presdate, Pressalolo, Sch 15719W, Trandate, Vescal
IUPAC-namn 2-hydroxy-5-[1-hydroxy-2-(4-phenylbutan-2-ylamino)ethyl]benzamide;hydrochloride
LEDER Cl.CC(CCc1ccccc1)NCC(O)c2ccc(O)c(c2)C(=O)N
Specifikationer
Kemiskt namn eller material Labetalol Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C19H24N2O3.ClH/c1-13(7-8-14-5-3-2-4-6-14)21-12-18(23)15-9-10-17(22)16(11-15)19(20)24;/h2-6,9-11,13,18,21-23H,7-8,12H2,1H3,(H2,20,24);1H
Formel vikt 364.1554