Beskrivning
Laboratory dishwasher with liquid dispensing and hot air drying DryPlus
- Delivery includes: Dosing module DOS K 85
- Performance/charge e.g. 128 laboratory flasks
- Efficient use of resources - variable speed heater pump
- Safety through monitoring - wash pressure and spray arm monitoring
- Efficient hot air drying - DryPlus
- Total security - link to process documentation possible
Equipment
- Outer casing stainless steel with 1 integrated dosing pump for oil applications and dosing module for liquid detergent
- circulation pump Qmax. 500l/min.
- circulation pump 0.8 kW
- TouchControl/15 programmes
- AutoClose - Automatic door lock
- Buzzer, acoustic signal at end of programme
- Waterproofsystem (WPS)
Specifikationer
Specifikationer
| Beskrivning | PG 8593 WW AD OIL Set DOS |
| Visa | Touch Screen |
| Produkttyp | Laboratory Glassware Washer |
| Certifikat och efterlevnad | EN 61010-1, EN 61010-2-40, EN 50581, CE Marked |
| Djup (metriskt) | 600 mm |
| Höjd (metrisk) | 835 mm |
| Bredd (metrisk) | 700 mm |
| Frekvens | 50 Hz |