Learn More
LACC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559081
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FGLKLNSQKN CHQTLLKTLN AVQYHHAAKA KFLCIMCCSN ISYERDGEQD NCEIETSNGL SALLEEFEIV SCPSMAATLY TIKQKIDEKN LSSIKVI Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 59%.
LACC1 Polyclonal antibody specifically detects LACC1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
LACC1 | |
Polyclonal | |
Unconjugated | |
LACC1 | |
9030625A04Rik; C12H13orf31; C13orf31; FAMIN; Fatty acid metabolism-immunity nexus; LACC1; laccase (multicopper oxidoreductase) domain containing 1; laccase domain containing 1; laccase domain-containing protein 1; purine nucleoside phosphorylase LACC1; laccase domain-containing protein 1; RGD1310185; si:ch211-187f22.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144811 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IV20 | |
LACC1 | |
Recombinant protein corresponding to Human LACC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only