Läs mer
Lacosamide, TRC
Produktkod. 30479452
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479452 10 mg
30470238 100 mg
30474357 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479452 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL09850010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 175481-36-4
Molekylformel C13 H18 N2 O3
Molekylvikt (g/mol) 250.29
Synonym (2R)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)-propanamide, Propanamide, 2-(acetylamino)-3-methoxy-N-(phenylmethyl)-, (R)-, (2R)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)propanamide, (2R)-2-Acetamido-N-benzyl-3-methoxypropanamide, (R)-2-Acetamido-N-benzyl-3-methoxypropionamide, ADD 243037, Erlosamide, Harkoseride, Lacosamide, Lacoset, Lacoset 50, SPM 927, Vimpat
IUPAC-namn (2R)-2-acetamido-N-benzyl-3-methoxypropanamide
LEDER COC[C@@H](NC(=O)C)C(=O)NCc1ccccc1
Specifikationer
Kemiskt namn eller material Lacosamide
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C13H18N2O3/c1-10(16)15-12(9-18-2)13(17)14-8-11-6-4-3-5-7-11/h3-7,12H,8-9H2,1-2H3,(H,14,17)(H,15,16)/t12-/m1/s1
Formel vikt 250.1317
Procent renhet >95%