Läs mer
Lacto-N-tetraose, TRC
Produktkod. 30484228
Change view
Kvantitet:
1 mg
2.5 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30484228 1 mg -
30468396 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30484228 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3002101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14116-68-8
Molekylformel C26 H45 N O21
Molekylvikt (g/mol) 707.63
Synonym R-lactotetraose-HSA, Beta-D-galacto-hexopyranosyl-(1→3)-2-acetamido-2-deoxy-beta-D-gluco-hexopyranosyl-(1→3)-beta-D-galacto-hexopyranosyl-(1→4)-D-gluco-hexopyranose, BetaD-Galp(1-3)betaD-GlcpNAc(1-3)betaD-Galp(1-4)D-Glcp
IUPAC-namn N-((2S,3R,4R,5R,6R)-2-(((2R,3S,4S,5R,6S)-3,5-dihydroxy-2-(hydroxymethyl)-6-(((2R,3S,4S,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)tetrahydro-2H-pyran-4-yl)oxy)-5-hydroxy-6-(hydroxymethyl)-4-(((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl)acetamide
LEDER CC(=O)N[C@H]1[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@H]3[C@@H](O)[C@@H](O)[C@H](O)O[C@@H]3CO)[C@@H]2O)O[C@H](CO)[C@H](O)[C@@H]1O[C@@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O
Specifikationer
Kemiskt namn eller material Lacto-N-tetraose
Kvantitet 1 mg
InChI formel InChI=1S/C26H45NO21/c1-6(32)27-11-21(47-25-18(39)15(36)12(33)7(2-28)44-25)13(34)8(3-29)43-24(11)48-22-14(35)9(4-30)45-26(19(22)40)46-20-10(5-31)42-23(41)17(38)16(20)37/h7-26,28-31,33-41H,2-5H2,1H3,(H,27,32)/t7-,8-,9-,10-,11-,12+,13+,14+,15+,16+,17-,18-,19-,20-,21-,22+,23-,24+,25+,26+/m1/s1
Formel vikt 707.2484